Learn More
BURKLE INC PTFE-valve, two-way, Ø 0.18 in. (4,5 mm), NW 0.08 in. (2 mm), autoklav.
Catalog No. 5000025542
Encompass_Preferred
Two-way valves made of solid PTFE with hose nozzles. Resistant to chemicals. Very easy connection thanks to its excellent antifriction properties. Blue handles made of PP.
Catalog No. 50-000-25542 Supplier BURKLE INC Supplier No. 86100010
Two-way valves made of solid PTFE with hose nozzles. Resistant to chemicals. Very easy connection thanks to its excellent antifriction properties. Blue handles made of PP.