Learn More
Milestone Inc PTFE vial 16/130
SDP
Catalog No. NC1628600
Encompass
PTFE vial 16/130 (Ø 16 mm/ 13 mm x 130mm) ca. 15 ml
Catalog No. NC1628600 Supplier Milestone Inc Supplier No. HB00086
PTFE vial 16/130 (Ø 16 mm/ 13 mm x 130mm) ca. 15 ml