Learn More
Analytical Sales & Services PTFE Washer for M4 Thumb Screw
Catalog No. 5000804942
Encompass
Replacement PTFE Washers for 96 Position Radiochemistry Reactor\n\n• Place between the Top Cover and the center Thumb Screw (TS350) to limit heat transfer to the Thumb Screw\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: 25\n\n \n\n 
Catalog No. 50-008-04942 Supplier Analytical Sales & Services Supplier No. WASH553
Replacement PTFE Washers for 96 Position Radiochemistry Reactor\n\n• Place between the Top Cover and the center Thumb Screw (TS350) to limit heat transfer to the Thumb Screw\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: 25\n\n \n\n 