Learn More
Perkin Elmer US LLC PTFE-Coated Torch Cassette O-Ring, 2.57 mm I.D. for Avio 200/500
Catalog No. 502113571
Encompass_Preferred
The O-ring is PTFE-coated, blue colored, has an inner diameter of 2.57 mm. Two of these o-rings are included in the Avio Adjustable Torch O-Ring Kit (N0790437)
Catalog No. 50-211-3571 Supplier Perkin Elmer US LLC Supplier No. N0791334
The O-ring is PTFE-coated, blue colored, has an inner diameter of 2.57 mm. Two of these o-rings are included in the Avio Adjustable Torch O-Ring Kit (N0790437)