Learn More
Perkin Elmer US LLC PTFE-Coated Torch Injector O-Ring, 8.74 mm I.D., for Avio 200/500
Catalog No. 502113583
Encompass_Preferred
This O-ring is included in the Injector Support Adapter for Cyclonic Spray Chambers (N0790141), Injector Support Adapter for Scott Spray Chamber (N0790156), Avio 200/500 O-Ring Kit for Scott Injector (N0790440), and the Avio 200/500 O-Ring Kit for Cyclonic Injector (N0790441).
Catalog No. 50-211-3583 Supplier Perkin Elmer US LLC Supplier No. N0791333
This O-ring is included in the Injector Support Adapter for Cyclonic Spray Chambers (N0790141), Injector Support Adapter for Scott Spray Chamber (N0790156), Avio 200/500 O-Ring Kit for Scott Injector (N0790440), and the Avio 200/500 O-Ring Kit for Cyclonic Injector (N0790441).