Learn More
Electron Microscopy Sciences These "PTFE" Printed Slides which are extremely hydrophobic and acetone proof, chemically resistant and autoclavable. The slides measure 3x1" (76x26x1mm) with clean wettable wells.
SDP
Catalog No. NC3395966
Encompass_Preferred
These "PTFE" Printed Slides which are extremely hydrophobic and acetone proof, chemically resistant and autoclavable. The slides measure 3x1" (76x26x1mm) with clean wettable wells.
Catalog No. NC3395966 Supplier Electron Microscopy Sciences Supplier No. 6341215
These "PTFE" Printed Slides which are extremely hydrophobic and acetone proof, chemically resistant and autoclavable. The slides measure 3x1" (76x26x1mm) with clean wettable wells.