Learn More
Vortex Sales Group PTG-M100 - Manual Powder Flowability and Angle of Repose Tester
Catalog No. NC3385242
Encompass_Preferred
PTG-M100 - Manual Powder Flowability and Angle of Repose Tester The PTG-M100 manual powder tester is used to measure the flow behavior of granules and powders in compliance with the EP <2.9.36>, EP <2.9.16>, USP <1174> Pharmacopoeia and ISO 4324 standards. This instrument is used to determine the powder flow time and the powder’s angle of repose. By using a laboratory balance also weight, density and the volume of the powder cone can be calculated. Furthermore, the EP “flowability” result can be calculated by measuring the flow time of 100g of sample through a specified pouring nozzle according to the flow through an orifice with funnel method
Catalog No. NC3385242 Supplier Vortex Sales Group Supplier No. PTGM100
PTG-M100 - Manual Powder Flowability and Angle of Repose Tester The PTG-M100 manual powder tester is used to measure the flow behavior of granules and powders in compliance with the EP <2.9.36>, EP <2.9.16>, USP <1174> Pharmacopoeia and ISO 4324 standards. This instrument is used to determine the powder flow time and the powder’s angle of repose. By using a laboratory balance also weight, density and the volume of the powder cone can be calculated. Furthermore, the EP “flowability” result can be calculated by measuring the flow time of 100g of sample through a specified pouring nozzle according to the flow through an orifice with funnel method