Learn More
Aviva Systems Biology Corp PTGDR Antibody - C-terminal region (ARP61296_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61296_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTGDR. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human PTGDR. Alternate names: AS1, ASRT1, DP, DP1, MGC49004, PTGDR1ApplicationWB. Species reactivity: Human
Catalog No. 50-287-333