Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci PTGDR2 Peptide
SDP

Catalog No. 16594665
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-594-665 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-594-665 Supplier ProSci Supplier No. 7191P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

PTGDR2 peptide is used for blocking the activity of PTGDR2 antibody (catalog number 7191P).

Specifications

Gene ID (Entrez) 11251
Species Human
Accession Number NP_004769
Common Name prostaglandin D2 receptor 2
Concentration 200 μg/mL
Content And Storage Store PTGDR2 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol PTGDR2
Quantity 0.05 mg
Sequence 13 amino acids near the carboxy terminus of human PTGDR2.
Synonym DP2, DL1R, CD294, CRTH2, GPR44, Prostaglandin D2 receptor 2, Chemoattractant receptor-homologous molecule expressed on Th2 cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.