Learn More
Aviva Systems Biology Corp PTGDS Antibody - N-terminal region (ARP48154_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48154_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTGDS. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PTGDS. Alternate names: PDS, PGD2, PGDS, PGDS2, LPGDS, L-PGDSApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59735