Learn More
Eurofins Discoverx ChemiBrite EP1 Prostanoid Receptor Stable Cell Line, 2 vials. (HTS099L)
Catalog No. 502005877
Encompass
The ChemiBrite EP1 Prostanoid Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin E2) based activation of PTGER1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTGER1, a naturally Gq coupled GPCR. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of PTGER1, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5877 Supplier Eurofins Discoverx Supplier No. HTS099L
Add to Cart
Add to Cart
Request a Quote
The ChemiBrite EP1 Prostanoid Receptor cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin E2) based activation of PTGER1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTGER1, a naturally Gq coupled GPCR. ChemiBrite cells also express a clytin variant, allowing for luminescent as well as fluorescent signal detection. Upon activation of PTGER1, the resulting calcium activity can be assayed in either detection mode. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.