Learn More
Aviva Systems Biology Corp PTGER3 Antibody - middle region (ARP34098_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34098_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTGER3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTGER3. Alternate names: EP3, EP3-I, EP3-II, EP3-III, EP3-IV, EP3e, MGC141828, MGC141829, MGC27302, PGE2-RApplicationWB. Species reactivity: Human
Catalog No. 50-014-55247