Learn More
Aviva Systems Biology Corp PTGES2 Antibody - middle region (ARP65698_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65698_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTGES2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human PTGES2. Alternate names: C9orf15, FLJ14038, GBF1, MGC11289, PGES2, mPGES-2, GBF-1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-288-544