Learn More
Aviva Systems Biology Corp PTGES3 Antibody - middle region (ARP58319_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58319_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTGES3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTGES3. Alternate names: P23, TEBP, cPGESApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53883