Learn More
Aviva Systems Biology Corp PTGES3 Antibody - middle region (ARP58321_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58321_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTGES3. It was validated on Western Blot. PTGES3,prostaglandin E synthase 3 (cytosolic),P23; TEBP; cPGES; PTGES3,prostaglandin E synthase 3,prostaglandin E synthase 3,Hsp90 co-chaperone,telomerase-binding protein p23,progesterone receptor complex p23,cytosolic prostaglandin E synthase,cytosolic prostaglandin E2 synthase,unactive progesterone receptor, 23 kDThe immunogen is a synthetic peptide directed towards the middle region of human PTGES3. Alternate names: P23, TEBP, cPGESApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-462