Learn More
Eurofins Discoverx ChemiScreen FP Prostanoid Receptor Stable Cell Line, 2 vials. (HTS093C)
Catalog No. 502005858
Encompass
The ChemiScreen FP Prostanoid Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin F2a) based activation of PTGFR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTGFR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of PTGFR, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5858 Supplier Eurofins Discoverx Supplier No. HTS093C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen FP Prostanoid Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin F2a) based activation of PTGFR GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTGFR alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of PTGFR, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.