Learn More
Eurofins Discoverx PathHunter PTGIR Total Internalization U2OS Cell Line, 2 vials. (93-0838C3)
Supplier:  Eurofins Discoverx 930838C3
Encompass
The PathHunter PTGIR Total Internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of PTGIR via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged PTGIR and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the PTGIR receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1394
Only null left
Add to Cart
Request a Quote