Learn More
Aviva Systems Biology Corp PTGIS Antibody - middle region (ARP44377_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44377_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTGIS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTGIS. Alternate names: CYP8, CYP8A1, MGC126858, MGC126860, PGIS, PTGIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-58795