Learn More
Bachem pTH (1-38) (human)
SDP
Supplier:  Bachem 4014853.0500
Encompass_Preferred
pTH (1-38) (human), H-Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-Val-Ala-Leu-Gly-OH, 78232-94-7, C197H319N59O55S2, Mr 4458.20, 0.5mg. Parathyroid Hormone (1-38), human.
Catalog No. 50-259-881