Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC pTH (1-38) (human) | 78232-94-7 | 98.8% | 4458.13 | 5 MG
SDP

Catalog No. 5000397576
Encompass_Preferred

pTH (1-38) (human) is a polypeptide that can be found by peptide screening. Peptide screening is a research tool that pools active peptides primarily by immunoassay. It can be used for protein interaction, functional analysis, and epitope screening, especially in the field of agent research and development.

  • Polypeptide for research applications
  • Supports protein interaction, functional analysis, and epitope screening
  • Molecular weight: 4458.13
  • High purity: 98.8%
  • Appearance: Solid, white to off-white
  • Soluble in DMSO at 50 mg/mL
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-97576 Supplier Medchemexpress LLC Supplier No. HYP48255MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

pTH (1-38) (human) is a polypeptide that can be found by peptide screening. Peptide screening is a research tool that pools active peptides primarily by immunoassay. It can be used for protein interaction, functional analysis, and epitope screening, especially in the field of agent research and development.

  • Polypeptide for research applications
  • Supports protein interaction, functional analysis, and epitope screening
  • Molecular weight: 4458.13
  • High purity: 98.8%
  • Appearance: Solid, white to off-white
  • Soluble in DMSO at 50 mg/mL
  • Recommended storage for powder: -80°C for 2 years or -20°C for 1 year
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.