Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pth (44-68) (human) (TFA) | 99.5% | 2836.08 (free base) | 5 MG
SDP

Catalog No. 5000396571
Encompass_Preferred

Pth (44-68) (human) (TFA) is a synthetic peptide, presented as a white to off-white solid, with a molecular weight of 2836.08 (free base). It boasts a high purity of 99.45% as determined by HPLC.

  • Peptide sequence: Arg-Asp-Ala-Gly-Ser-Gln-Arg-Pro-Arg-Lys-Lys-Glu-Asp-Asn-Val-Leu-Val-Glu-Ser-His-Glu-Lys-Ser-Leu-Gly (TFA salt)
  • High purity (99.45% by HPLC)
  • Appearance: white to off-white solid
  • Stable storage conditions: powder at -80°C for 2 years or -20°C for 1 year; in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-96571 Supplier Medchemexpress LLC Supplier No. HYP4697A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pth (44-68) (human) (TFA) is a synthetic peptide, presented as a white to off-white solid, with a molecular weight of 2836.08 (free base). It boasts a high purity of 99.45% as determined by HPLC.

  • Peptide sequence: Arg-Asp-Ala-Gly-Ser-Gln-Arg-Pro-Arg-Lys-Lys-Glu-Asp-Asn-Val-Leu-Val-Glu-Ser-His-Glu-Lys-Ser-Leu-Gly (TFA salt)
  • High purity (99.45% by HPLC)
  • Appearance: white to off-white solid
  • Stable storage conditions: powder at -80°C for 2 years or -20°C for 1 year; in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.