Learn More
FUJIFILM Biosciences PTH-amino Acids Mobile Phase A (for Gradient Elution), 1L
Catalog No. NC1465462
Encompass_Preferred
This product is for research use only. Do not administer it to human. The Edman method developed by Edman is widely used as an extremely excellent method to determine the amino acid sequence of proteins and peptides. Since Edman and Begg developed the Automatic Edman method, it is required to reduce and speed up the analysis, and analyzes on the order of pmol are carried out using sequencers and the like. This product is eluent for Shimadzu Automatic Protein Primary Structure Analyzer (Gradient System).
Catalog No. NC1465462 Supplier FUJIFILM Biosciences Supplier No. 16328261
This product is for research use only. Do not administer it to human. The Edman method developed by Edman is widely used as an extremely excellent method to determine the amino acid sequence of proteins and peptides. Since Edman and Begg developed the Automatic Edman method, it is required to reduce and speed up the analysis, and analyzes on the order of pmol are carried out using sequencers and the like. This product is eluent for Shimadzu Automatic Protein Primary Structure Analyzer (Gradient System).