Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5741MSM6P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 3H9 + PTH/1175. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: Human parathyroid gland carcinoma. Cellular Staining: Cytoplasmic, Secreted. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Synthetic peptide corresponding to amino acids 1 to 34 of PTH conjugated to a carrier (3H9); Recombinant fragment (84 amino acid residues from C-terminus) of human PTH protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 9kDa.Chromosome: 11p15.3-p15.1. Gene Symbol: PTH. Gene ID: 5741. Ensemble: ENSG00000152266. Uniprot: NP_001303281.
Catalog No. 50-188-5094