Learn More
Genetex Inc Parathyroid Hormone Monoclonal Antibody [MSVA-525R] HistoMAX™ (GTX04405) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04405
Encompass_Preferred
The Parathyroid Hormone antibody [MSVA-525R] HistoMAX™ (GTX04405) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human kidney tissue using GTX04405 Parathyroid Hormone antibody [MSVA-525R] HistoMAX. Parathyroid hormone immunostaining is lacking in the kidney.
Catalog No. NC3666775