Learn More
Eurofins Discoverx PathHunter PTH Bioassay Kit, 2-Plate. (93-0315Y2-00047)
Catalog No. 502000257
Encompass
The PathHunter PTH Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses PTH based activation of PTHR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTHR and EA-tagged β-Arrestin-2. Activation of PTHR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-0257 Supplier Eurofins Discoverx Supplier No. 930315Y200047
The PathHunter PTH Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses PTH based activation of PTHR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTHR and EA-tagged β-Arrestin-2. Activation of PTHR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.