Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PTH Human 100ug
SDP

Catalog No. 5000196686
Encompass_Preferred

Recombinant full-length human parathyroid hormone supplied as a lyophilized, tag-free protein for research applications involving mineral ion metabolism, bone remodeling, receptor binding, and signaling assays. The product is characterized by a defined amino acid sequence, high purity, and recommended storage and reconstitution conditions for reliable experimental use.

  • Full-length human parathyroid hormone protein.
  • Tag-free recombinant protein expressed in E. coli.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 9-15 kDa.
  • Lyophilized formulation with validated buffer options for stability.
  • Stable at -20°C for long-term storage; short-term stability at 4°C after reconstitution.
  • Supplied in multiple pack sizes to suit experimental scale.

Catalog No. 50-001-96686 Supplier Medchemexpress LLC Supplier No. HYP7112100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human parathyroid hormone supplied as a lyophilized, tag-free protein for research applications involving mineral ion metabolism, bone remodeling, receptor binding, and signaling assays. The product is characterized by a defined amino acid sequence, high purity, and recommended storage and reconstitution conditions for reliable experimental use.

  • Full-length human parathyroid hormone protein.
  • Tag-free recombinant protein expressed in E. coli.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 9-15 kDa.
  • Lyophilized formulation with validated buffer options for stability.
  • Stable at -20°C for long-term storage; short-term stability at 4°C after reconstitution.
  • Supplied in multiple pack sizes to suit experimental scale.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.