Recombinant human parathyroid hormone (PTH) is a biologically active protein that regulates calcium and phosphorus homeostasis by acting on bone, kidney, and intestine. Supplied lyophilized in microgram-scale quantities for research applications, handle and reconstitute according to manufacturer guidance to preserve activity and stability.
- High purity greater than 95% as determined by reducing SDS-PAGE.
- Biologically active recombinant human sequence relevant to calcium and phosphate regulation.
- Tag free formulation to minimize extraneous sequences and downstream interference.
- Available in microgram-scale lyophilized quantities for assay flexibility.
- Recommended storage lyophilized at -20°C; aliquot and freeze at -20°C or -80°C after reconstitution for extended storage.
- Reconstitution guidance provided; avoid preparing solutions below 100 μg/mL in ddH2O.