Learn More
Echelon Biosciences Research Labs PTH Related Peptide [Leu11, DTrp12] (7-34) amide, human - 1 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 231961MG
Encompass_Preferred
PTH Related Peptide [Leu11, DTrp12] (7-34) amide, human - 1mg , Leu-Leu-His-Asp-Leu-DTrp-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Phe-Phe-Leu-His-His-Leu-Ile-Ala-Glu-Ile-His-Thr-Ala-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8306