Learn More
Echelon Biosciences Research Labs PTH [Nle8,18,Tyr34] (3-34), amide, bovine; Parathyroid Hormone - 0.5 mg, CAS#: 64763-77-5
SDP
Supplier:  Echelon Biosciences Research Labs 233350.5MG
Encompass_Preferred
PTH [Nle8,18,Tyr34] (3-34), amide, bovine; Parathyroid Hormone - 0.5mg , Ser-Glu-Ile-Gln-Phe-Nle-His-Asn-Leu-Gly-Lys-His-Leu-Ser-Ser-Nle-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Tyr-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8309