Learn More
Drg International Inc Pth (Parathyroid) Intact Elisa
Supplier:  Drg International Inc EIA3645
Encompass_Preferred
An enzyme immunoassay for the quantitative determination of Intact-PTH (Parathyroid Hormone) in serum. PTH (Parathyroid hormone, Parathormone, Parathyrin) is biosynthesized in the parathyroid gland as a pre-proparathyroid hormone. In subjects with renal insufficiency, C-terminal and midregion PTH assays typically give elevated PTH results, as reflected by impaired renal clearance. The DRG Intact PTH Immunoassay is a two-site ELISA [Enzyme-Linked ImmunoSorbent Assay] for the measurement of the biologically intact 84 amino acid chain of PTH. One antibody is prepared to bind only the mid-region and C-terminal PTH 39-84 and this antibody is biotinylated. The other antibody is prepared to bind only the N-terminal PTH 1-34 and this antibody is labeled with horseradish peroxidase [HRP] for detection. Although mid-region and C-terminal fragments are bound by the biotinylated anti-PTH (39-84), only the intact PTH 1-84 forms the sandwich complex necessary for detection.
Catalog No. 50-194-7596