Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10097MA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His. Host: Mammal. Species Reactivity: Pig. Protein Expression: A DNA sequence encoding the Sus scrofa (Pig) Parathyroid Hormone, was expressed in the hosts and tags indicated. AA Sequence: MTKEEQIFLL HRAQAQCEKR LKEVLQRPAD IMESDKGWAS APTSGKPRKE KASGKLYPES GEDTGSRHQG RPCLPEWDHI LCWP. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: PTH. Gene ID: 397675. Uniprot: P50133.
Catalog No. 50-188-5495