Learn More
Bachem pTH-Related Protein (1-34) (human, mouse, rat)
SDP
Supplier:  Bachem 4017147.0500
Encompass_Preferred
pTH-Related Protein (1-34) (human, mouse, rat) , H-Ala-Val-Ser-Glu-His-Gln-Leu-Leu-His-Asp-Lys-Gly-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Phe-Phe-Leu-His-His-Leu-Ile-Ala-Glu-Ile-His-Thr-Ala-OH, 112540-82-6, C180H287N57O48, Mr 4017.61, 0.5mg. Hypercalcemia of Malignancy Factor (1-34) (human, mouse, rat), pTHrP (1-34) (human, mouse, rat). V.Paspaliaris et al. showed that daily administration of pTHrP (1-34) to rats was anabolic on bone by increasing bone formation. This treatment could inhibit the rapid decline in bone formation due to denervation.
Catalog No. 50-260-109