Learn More
Sino Biological PTH1R / PTHR1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12232RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PTH1R / PTHR1 (rh PTH1R / PTHR1; Catalog#12232-H08H; Q03431; Met 1-Gly 188). PTH1R / PTHR1 specific IgG was purified by Human PTH1R / PTHR1 affinity chromatography.
Catalog No. 50-224-8677