Learn More
Sino Biological PTH1R / PTHR1 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 12232R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PTH1R / PTHR1 (rh PTH1R / PTHR1; Catalog#12232-H08H; Q03431; Met1-Gly188).
Catalog No. 50-224-7903