Learn More
ACROBiosystems Recombinant Protein;Human PTH1R / PTHR1 Protein;HEK293;1MG;PTR,PTH1R,PTHR,PTHR1,PTH/PTHr receptor,PTH1 receptor
Supplier:  ACROBiosystems PTRH52221MG
Encompass_Preferred
Recombinant Protein;Human PTH1R / PTHR1 Protein;1MG;Human PTH1R, His Tag (PTR-H5222) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Gly 188 (Accession # NP_000307).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 23-40 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-165-8363