Learn More
Aviva Systems Biology Corp PTH2R Antibody - C-terminal region (ARP64926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTH2R. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human PTH2R. Alternate names: PTHR2ApplicationWB. Species reactivity: Cow, Guinea Pig, Human
Catalog No. 50-288-394