Learn More
Aviva Systems Biology Corp PTHLH Antibody - middle region (ARP33885_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33885_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PTHLH. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PTHLH. Alternate names: HHM, PLP, BDE2, PTHR, PTHRPApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55187