Learn More
Eurofins Discoverx ChemiScreen PTH1 Parathyroid Hormone Receptor Stable Cell Line, 2 vials. (HTS030C)
Catalog No. 502005703
Encompass
The ChemiScreen PTH1 Parathyroid Hormone Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. parathyroid hormone) based activation of PTH1R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTH1R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of PTH1R, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5703 Supplier Eurofins Discoverx Supplier No. HTS030C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen PTH1 Parathyroid Hormone Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. parathyroid hormone) based activation of PTH1R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses PTH1R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of PTH1R, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.