Learn More
Cayman Chemical PTI-125 (hydrochloride), Purity: ≥98%, CAS: 2480226-06-8, 100 mg
SDP
Catalog No. 502943731
Encompass_Preferred
A purported binding agent of filamin A; the data regarding PTI-125 have prompted expressions of concern by the journals in which the data were published
Catalog No. 50-294-3731 Supplier Cayman Chemical Supplier No. 41859100 MG
A purported binding agent of filamin A; the data regarding PTI-125 have prompted expressions of concern by the journals in which the data were published