Learn More
Cayman Chemical PTI-1 (hydrochloride), Purity: ≥95%, CAS: 1400742-46-2, 10 mg
SDP
Catalog No. 502894454
Encompass_Preferred
A synthetic cannabinoid (CB) that contains the 1-pentyl-indole structure found in potent agonists of the central cannabinoid (CB1) receptor, like JWH 018 (Item No. 10900), linked to a thiazole-based side chain; intended for forensic and research applications
Catalog No. 50-289-4454 Supplier Cayman Chemical Supplier No. 900194810 MG
A synthetic cannabinoid (CB) that contains the 1-pentyl-indole structure found in potent agonists of the central cannabinoid (CB1) receptor, like JWH 018 (Item No. 10900), linked to a thiazole-based side chain; intended for forensic and research applications