Learn More
Aviva Systems Biology Corp PTK2B Antibody - C-terminal region (ARP32684_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32684_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PTK2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PTK2B. Alternate names: PKB, PTK, CAKB, FAK2, PYK2, CADTK, FADK2, RAFTKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54706