Learn More
Aviva Systems Biology Corp PTK6 Antibody - middle region (ARP56657_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56657_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PTK6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PTK6. Alternate names: BRK, FLJ42088ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-61942