Learn More
Cayman Chemical PTK7 EXrAClular DomaInhu 100ug
SDP
Catalog No. 502944096
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged PTK7 extracellular domain expressed in HEK293 cells • Amino acids: 1-704 • MW: 76.1 kDa
Catalog No. 50-294-4096 Supplier Cayman Chemical Supplier No. 40523100 UG
Source: Recombinant human C-terminal His-tagged PTK7 extracellular domain expressed in HEK293 cells • Amino acids: 1-704 • MW: 76.1 kDa