Learn More
Sino Biological Recombinant Human PSKH1 Protein (GST Tag)
Catalog No. 502573509
Encompass_Preferred
Recombinant human PSKH1 (2-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3509 Supplier Sino Biological Supplier No. P8934G100
Recombinant human PSKH1 (2-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.