Learn More
Sino Biological Recombinant Human PTK2/FAK Protein (Full Length, GST Tag), Active
Catalog No. 502573505
Encompass_Preferred
Recombinant full length human FAK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3505 Supplier Sino Biological Supplier No. P9110G50
Recombinant full length human FAK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.