Learn More
BEZWADA BIOMEDICAL LLC Poly(trimethylene carbonate-co-L-lactide), TMC:L-lactide 90:10, viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Catalog No. 501904495
Encompass_Preferred
Polytrimethylene carbonate amorphous rubbery polymer. Copolymers of Trimethylene carbonate is prepared for varying absorption profile, stiffness and crsytallinity. The polymer is elastomeric. Polymers are used in drug delivery and medical device coatings.
Catalog No. 50-190-4495 Supplier BEZWADA BIOMEDICAL LLC Supplier No. 5391
Polytrimethylene carbonate amorphous rubbery polymer. Copolymers of Trimethylene carbonate is prepared for varying absorption profile, stiffness and crsytallinity. The polymer is elastomeric. Polymers are used in drug delivery and medical device coatings.