Learn More
Cell Signaling Technology PTMScan™ Phospho-Enrichment IMAC Fe-NTA Magnetic Beads, 1000 μL
SDP
Numéro de catalogue. NC2469849
Encompass
PTMScan™ Phospho-Enrichment IMAC Fe-NTA Magnetic Beads utilize immobilized metal affinity chromatography to capture phosphorylated peptides for analysis. Negatively charged phosphate groups bind to the positively charged metal ions on the magnetic beads. This technology enables the isolation, identification, and quantitation of phosphorylated cellular peptides via liquid chromatography tandem mass spectrometry (LC-MS/MS), providing a comprehensive overview of the phosphoproteome in tissue and cell samples.
  • Efficient tool for phosphopeptide enrichment with little to no bias for phospho-residue context.
  • Superior selectivity for phosphopeptides compared to iminodiacetic acid (IDA) based systems.
  • Compatible with independent use or as a complement to immunoaffinity-based enrichment workflows.
  • Supplied in 20% ethanol for stability.
  • Bead diameter of 50 microns.
Numéro de catalogue. NC2469849 Fournisseur Cell Signaling Technology Code fournisseur 20432L
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
PTMScan™ Phospho-Enrichment IMAC Fe-NTA Magnetic Beads utilize immobilized metal affinity chromatography to capture phosphorylated peptides for analysis. Negatively charged phosphate groups bind to the positively charged metal ions on the magnetic beads. This technology enables the isolation, identification, and quantitation of phosphorylated cellular peptides via liquid chromatography tandem mass spectrometry (LC-MS/MS), providing a comprehensive overview of the phosphoproteome in tissue and cell samples.
  • Efficient tool for phosphopeptide enrichment with little to no bias for phospho-residue context.
  • Superior selectivity for phosphopeptides compared to iminodiacetic acid (IDA) based systems.
  • Compatible with independent use or as a complement to immunoaffinity-based enrichment workflows.
  • Supplied in 20% ethanol for stability.
  • Bead diameter of 50 microns.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.