Learn More
Cell Signaling Technology PTMScan™ O-GlcNAc [GlcNAc-S/T] Motif Kit, 10 Assays
SDP
Catalog No. 503131641
Encompass
PTMScan™ technology employs a proprietary methodology for post-translationally modified peptide enrichment by immunoprecipitation using a specific agarose bead-conjugated antibody in conjunction with liquid chromatography-tandem mass spectrometry (LC-MS/MS) for quantitative profiling of O-GlcNAc modified sites in cellular proteins. The workflow provides a global, unbiased overview of O-GlcNAc modifications across cell and tissue samples of any species, enabling researchers to isolate, identify, and quantify large numbers of modified peptides.
  • Enables quantitative profiling of O-GlcNAc [GlcNAc-S/T] motifs.
  • Utilizes high-affinity, agarose bead-conjugated antibodies.
  • Compatible with diverse cell and tissue sample types.
  • Designed for 10 assays.
  • Validated for use in proteomics research applications.
  • Supplied with immunoaffinity beads and 10X IAP buffer.
Catalog No. 50-313-1641 Supplier Cell Signaling Technology Supplier No. 95220S
PTMScan™ technology employs a proprietary methodology for post-translationally modified peptide enrichment by immunoprecipitation using a specific agarose bead-conjugated antibody in conjunction with liquid chromatography-tandem mass spectrometry (LC-MS/MS) for quantitative profiling of O-GlcNAc modified sites in cellular proteins. The workflow provides a global, unbiased overview of O-GlcNAc modifications across cell and tissue samples of any species, enabling researchers to isolate, identify, and quantify large numbers of modified peptides.
  • Enables quantitative profiling of O-GlcNAc [GlcNAc-S/T] motifs.
  • Utilizes high-affinity, agarose bead-conjugated antibodies.
  • Compatible with diverse cell and tissue sample types.
  • Designed for 10 assays.
  • Validated for use in proteomics research applications.
  • Supplied with immunoaffinity beads and 10X IAP buffer.