Learn More
Cell Signaling Technology PTMScan™ Pan-Methyl Lysine Kit, 10 Assays, 1 Kit
SDP
Catalog No. 503132154
Encompass
The PTMScan™ Pan-Methyl Lysine Kit utilizes a proprietary methodology for the enrichment of post-translationally modified peptides. By employing specific bead-conjugated antibodies and liquid chromatography-tandem mass spectrometry (LC-MS/MS), this kit allows for the quantitative profiling of methyl-lysine sites in cellular proteins. It facilitates the unbiased, global isolation and identification of modified peptides across cell and tissue samples from any species.
  • Enables quantitative profiling of methyl-lysine modification sites
  • Supports unbiased global analysis of post-translational modifications
  • Includes immunoaffinity beads and 10X IAP buffer
  • Suitable for applications involving cellular protein enrichment
  • Designed for research use only
Catalog No. 50-313-2154 Supplier Cell Signaling Technology Supplier No. 14809S
The PTMScan™ Pan-Methyl Lysine Kit utilizes a proprietary methodology for the enrichment of post-translationally modified peptides. By employing specific bead-conjugated antibodies and liquid chromatography-tandem mass spectrometry (LC-MS/MS), this kit allows for the quantitative profiling of methyl-lysine sites in cellular proteins. It facilitates the unbiased, global isolation and identification of modified peptides across cell and tissue samples from any species.
  • Enables quantitative profiling of methyl-lysine modification sites
  • Supports unbiased global analysis of post-translational modifications
  • Includes immunoaffinity beads and 10X IAP buffer
  • Suitable for applications involving cellular protein enrichment
  • Designed for research use only