Learn More
Cell Signaling Technology PTMScan™ Phospho-Enrichment IMAC Fe-NTA Magnetic Beads, 200 μL
SDP
Catalog No. 502050586
Encompass
PTMScan™ Phospho-Enrichment IMAC Fe-NTA Magnetic Beads utilize immobilized metal affinity chromatography to capture phosphorylated peptides from complex biological samples. By leveraging the interaction between negatively charged phosphate groups and positively charged Fe3+ ions, these beads enable specific and sensitive enrichment for global phosphoproteome analysis in combination with liquid chromatography tandem mass spectrometry (LC-MS/MS).
  • Provides efficient phosphopeptide enrichment with minimal bias for phospho-residue context.
  • Features 50 μm diameter magnetic beads for streamlined handling and separation.
  • Supplied in 20% ethanol for stability.
  • Includes a detailed protocol for cell lysis, peptide digestion, purification, and enrichment.
  • Intended for research use only.
Catalog No. 50-205-0586 Supplier Cell Signaling Technology Supplier No. 20432S
PTMScan™ Phospho-Enrichment IMAC Fe-NTA Magnetic Beads utilize immobilized metal affinity chromatography to capture phosphorylated peptides from complex biological samples. By leveraging the interaction between negatively charged phosphate groups and positively charged Fe3+ ions, these beads enable specific and sensitive enrichment for global phosphoproteome analysis in combination with liquid chromatography tandem mass spectrometry (LC-MS/MS).
  • Provides efficient phosphopeptide enrichment with minimal bias for phospho-residue context.
  • Features 50 μm diameter magnetic beads for streamlined handling and separation.
  • Supplied in 20% ethanol for stability.
  • Includes a detailed protocol for cell lysis, peptide digestion, purification, and enrichment.
  • Intended for research use only.